Learn More
TARGETMOL CHEMICALS INC Acyclovir 200MG
SDP
Catalog No. 502880466
Encompass_Preferred
Also available in 1 mL, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Acyclovir (Aciclovir) is a synthetic analog of the purine nucleoside, guanosine, with potent antiviral activity against herpes simplex viruses type 1 and 2, varicella-zoster virus and other viruses of the herpesvirus family. Purity 100%
Catalog No. 50-288-0466 Supplier TARGETMOL CHEMICALS INC Supplier No. T1454200MG
Also available in 1 mL, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Acyclovir (Aciclovir) is a synthetic analog of the purine nucleoside, guanosine, with potent antiviral activity against herpes simplex viruses type 1 and 2, varicella-zoster virus and other viruses of the herpesvirus family. Purity 100%