Learn More
Medchemexpress LLC Acyclovir-d4 | 1185179-33-2 | 98.66% | 229.23 | 1 MG
SDP
Catalog No. 5000385034
Encompass_Preferred
Acyclovir-d4 is the deuterium labeled Acyclovir. Acyclovir is a guanosine analogue and an orally active antiviral agent. It inhibits HSV-1, HSV-2, and varicella-zoster virus, interfering with viral DNA polymerization.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-85034 Supplier Medchemexpress LLC Supplier No. HY17422S11MG
Acyclovir-d4 is the deuterium labeled Acyclovir. Acyclovir is a guanosine analogue and an orally active antiviral agent. It inhibits HSV-1, HSV-2, and varicella-zoster virus, interfering with viral DNA polymerization.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.