Learn More
Medchemexpress LLC Acyclovir triphosphate sodium | 221119-43-3 | 99.7% | C8H14N5O12P3.xNa | 25 MG
SDP
Catalog No. 5000354665
Encompass_Preferred
Acyclovir triphosphate sodium (AcycloGTP sodium) is a derivative of Acyclovir that functions as an inhibitor of viral DNA polymerase by acting as an analog to deoxyguanosine triphosphate (dGTP). This compound is also recognized as an inhibitor of HIV-1 reverse transcriptase, ultimately leading to the termination of viral DNA synthesis. It is intended for research use only.
  • Competitively inhibits viral DNA polymerase.
  • Acts as an analog to deoxyguanosine triphosphate (dGTP).
  • Inhibits HIV-1 reverse transcriptase.
  • Causes termination of viral DNA synthesis.
  • Available for research purposes.
Catalog No. 50-003-54665 Supplier Medchemexpress LLC Supplier No. 25MGHY142028A
Acyclovir triphosphate sodium (AcycloGTP sodium) is a derivative of Acyclovir that functions as an inhibitor of viral DNA polymerase by acting as an analog to deoxyguanosine triphosphate (dGTP). This compound is also recognized as an inhibitor of HIV-1 reverse transcriptase, ultimately leading to the termination of viral DNA synthesis. It is intended for research use only.
  • Competitively inhibits viral DNA polymerase.
  • Acts as an analog to deoxyguanosine triphosphate (dGTP).
  • Inhibits HIV-1 reverse transcriptase.
  • Causes termination of viral DNA synthesis.
  • Available for research purposes.