Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Acyclovir triphosphate sodium | 221119-43-3 | MFCD09970911 | 99.7% | C8H14N5O12P3.xNa | 5 MG
SDP

Catalog No. 5000221655
Encompass_Preferred

Acyclovir triphosphate sodium is the sodium salt of acyclovir's active triphosphate metabolite, supplied as a high-purity nucleotide analog for biochemical research. It acts as a chain-terminating inhibitor of viral polymerases and is used in enzymatic assays and nucleic acid studies.

  • High purity (HPLC 99.7%).
  • Molecular formula C8H14N5O12P3·xNa.
  • Functions as a nucleotide analog and polymerase inhibitor.
  • Provided in research-scale quantities and solution formats (e.g., 5 mg; 100 mM in water).
  • Intended for biochemical assays, polymerase inhibition studies, and nucleic acid research.
  • Typically stored frozen at -20 °C per supplier guidance.

Catalog No. 50-002-21655 Supplier Medchemexpress LLC Supplier No. HY142028A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Acyclovir triphosphate sodium is the sodium salt of acyclovir's active triphosphate metabolite, supplied as a high-purity nucleotide analog for biochemical research. It acts as a chain-terminating inhibitor of viral polymerases and is used in enzymatic assays and nucleic acid studies.

  • High purity (HPLC 99.7%).
  • Molecular formula C8H14N5O12P3·xNa.
  • Functions as a nucleotide analog and polymerase inhibitor.
  • Provided in research-scale quantities and solution formats (e.g., 5 mg; 100 mM in water).
  • Intended for biochemical assays, polymerase inhibition studies, and nucleic acid research.
  • Typically stored frozen at -20 °C per supplier guidance.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.