Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acyclovir, USP, 98-101%, Spectrum™ Chemical
SDP

Catalog No. 18600318 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
1 kg
12 kg
Packaging:
Amber Glass Bottle
Poly Bottle
Poly Pail
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-600-318 500 g Poly Bottle
18-600-315 5 g Amber Glass Bottle
18-600-316 25 g Poly Bottle
18-600-317 100 g Poly Bottle
18-600-319 1 kg Poly Pail
18-600-320 12 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18-600-318 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. A1816500GM
Only null left
Add to Cart
Add to Cart

A1816, 59277-89-3, C8H11N5O3

Spectrum™ Chemical Acyclovir, USP is the most commonly used antiviral drug and is on the World Health Organization′s list of essential medicines. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 59277-89-3
Molecular Formula C8H11N5O3
Molecular Weight (g/mol) 225.21
MDL Number MFCD00057880
InChI Key MKUXAQIIEYXACX-UHFFFAOYSA-N
IUPAC Name 2-amino-9-[(2-hydroxyethoxy)methyl]-6,9-dihydro-1H-purin-6-one
SMILES NC1=NC2=C(N=CN2COCCO)C(=O)N1

Specifications

CAS Max % 100%
Packaging Poly Bottle
Quantity 500 g
Percent Purity 98 to 101%
Grade USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.