Learn More
Abcam Acyclovir, viral DNA polymerase inhibitor, 100MG
Catalog No. 502707409
Encompass_Preferred
MW 225.21 g/mol, Purity >99%. Guanosine derivative, competitively inhibits viral DNA polymerase. High specificity for herpes simplex 1 and 2 (EC₅₀ values are 0.04 and 0.44 μM, respectively) and varicella-zoster viruses (IC₅₀ = 0.22 μM).
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7409 Supplier Abcam Supplier No. AB120631100MG
MW 225.21 g/mol, Purity >99%. Guanosine derivative, competitively inhibits viral DNA polymerase. High specificity for herpes simplex 1 and 2 (EC₅₀ values are 0.04 and 0.44 μM, respectively) and varicella-zoster viruses (IC₅₀ = 0.22 μM).
The product is subject to the following: Abcam Restricted Use Statement