Learn More
Medchemexpress LLC Acyl coenzyme A synthetase (ACS) | 9013-18-7 | 100 U
SDP
Catalog No. 5000398702
Encompass_Preferred
Acyl coenzyme A synthetase (ACS) is frequently utilized in biochemical research. It facilitates the activation of fatty acids by coenzyme A through a two-step thioesterification, leading to the production of acyl coenzyme A. This compound then participates in various anabolic and catabolic lipid metabolism pathways, as well as the TCA cycle in aerobic respiration.
  • Used in biochemical research.
  • Catalyzes the activation of fatty acids by coenzyme A.
  • Involved in anabolic and catabolic lipid metabolism pathways.
  • Participates in the TCA cycle in aerobic respiration.
  • Activity loss of solid powder when stored at 37°C for 7 days is less than 10%.
  • Can be dissolved in water at a concentration of 1 mg/mL, or in buffer according to specific experimental references.
Catalog No. 50-003-98702 Supplier Medchemexpress LLC Supplier No. HYP2832100U
Acyl coenzyme A synthetase (ACS) is frequently utilized in biochemical research. It facilitates the activation of fatty acids by coenzyme A through a two-step thioesterification, leading to the production of acyl coenzyme A. This compound then participates in various anabolic and catabolic lipid metabolism pathways, as well as the TCA cycle in aerobic respiration.
  • Used in biochemical research.
  • Catalyzes the activation of fatty acids by coenzyme A.
  • Involved in anabolic and catabolic lipid metabolism pathways.
  • Participates in the TCA cycle in aerobic respiration.
  • Activity loss of solid powder when stored at 37°C for 7 days is less than 10%.
  • Can be dissolved in water at a concentration of 1 mg/mL, or in buffer according to specific experimental references.