Learn More
Cayman Chemical Acylated Ghrelin human Expre
SDP
Catalog No. 502886814
Encompass_Preferred
This EIA kit specifically measures the acylated form of ghrelin.
Catalog No. 50-288-6814 Supplier Cayman Chemical Supplier No. 100063061 EA
This EIA kit specifically measures the acylated form of ghrelin.