Learn More
Cayman Chemical Acylated Ghrelin mouse rat
SDP
Catalog No. 502887019
Encompass_Preferred
This EIA kit specifically measures the acylated form of ghrelin.
Catalog No. 50-288-7019 Supplier Cayman Chemical Supplier No. 100063071 EA
This EIA kit specifically measures the acylated form of ghrelin.