Learn More
Cayman Chemical AcYVADpNAtrIfluoroACTAte 10mg
SDP
Catalog No. 502965451
Encompass_Preferred
A colorimetric substrate whose amino acids YVAD have been shown to be a preferred cleavage site for caspase-1 and -4
Catalog No. 50-296-5451 Supplier Cayman Chemical Supplier No. 1748710 MG
A colorimetric substrate whose amino acids YVAD have been shown to be a preferred cleavage site for caspase-1 and -4