Learn More
Medchemexpress LLC AD 01 | 959961-23-0 | 99.98% | 2703.94 | 1 MG
SDP
Catalog No. 5000423535
Encompass_Preferred
AD 01 is a 24 amino acid peptide derived from FKBPL (FK506-binding protein like) with potent anti-angiogenic activity. It binds to the CD44 receptor, inhibiting tumor cell migration in a CD44-dependent manner. In vitro studies show AD-01 up-regulates CD44 expression in MDA-MB-231 and HMEC-1 cells. In vivo, AD 01 has demonstrated significant anti-tumor activity in DU145 tumors in Balb-c severe compromised immuno-deficient (SCID) mice, significantly lowering tumor size.
  • 24 amino acid peptide
  • Potent anti-angiogenic activity
  • Binds to the CD44 receptor
  • Inhibits tumor cell migration in a CD44-dependent manner
  • Up-regulates CD44 expression in MDA-MB-231 and HMEC-1 cells in vitro
  • Demonstrates significant anti-tumor activity in DU145 tumors in vivo
  • Significantly lowers tumor size in vivo
Catalog No. 50-004-23535 Supplier Medchemexpress LLC Supplier No. HYP22841MG
AD 01 is a 24 amino acid peptide derived from FKBPL (FK506-binding protein like) with potent anti-angiogenic activity. It binds to the CD44 receptor, inhibiting tumor cell migration in a CD44-dependent manner. In vitro studies show AD-01 up-regulates CD44 expression in MDA-MB-231 and HMEC-1 cells. In vivo, AD 01 has demonstrated significant anti-tumor activity in DU145 tumors in Balb-c severe compromised immuno-deficient (SCID) mice, significantly lowering tumor size.
  • 24 amino acid peptide
  • Potent anti-angiogenic activity
  • Binds to the CD44 receptor
  • Inhibits tumor cell migration in a CD44-dependent manner
  • Up-regulates CD44 expression in MDA-MB-231 and HMEC-1 cells in vitro
  • Demonstrates significant anti-tumor activity in DU145 tumors in vivo
  • Significantly lowers tumor size in vivo