Learn More
Euthanex AD-5000 Digital Anesthesia System, including TEC 3 Vaporizer, E-Z Heat Surgical Beds Package, Touch-Screen, Regulator, Induction Chamber and Charcoal Filters
Catalog No. NC1880364
Encompass_Preferred
The AD-5000 Anesthesia System advances anesthesia into the digital era. The AD-5000 uses the always reliable TEC 3 vaporizer as the cornerstone and enhances its features with a digital interface. The system incorporates a vivid touch screen that allows the operator to have a full range of features, including a precise range of 0.5 to 2.5 LPM flow rate per connection, choice of 2-3 breathing circuits and induction chamber, user choice of multiple outputs while automatically adjusting for the correct flow rates. The induction chamber incorporates an automatic flush button to prevent undesired exposure when transferring animals. Includes individual timers for accurate monitoring and prevention of over exposure to the patients. Also includes all new digital warming beds to maintain body temperature during procedures, for both the induction chamber and breathing circuits between 95 to 104 degrees Fahrenheit.
Catalog No. NC1880364 Supplier Euthanex Supplier No. AD5000
Add to Cart
Add to Cart
Request a Quote
The AD-5000 Anesthesia System advances anesthesia into the digital era. The AD-5000 uses the always reliable TEC 3 vaporizer as the cornerstone and enhances its features with a digital interface. The system incorporates a vivid touch screen that allows the operator to have a full range of features, including a precise range of 0.5 to 2.5 LPM flow rate per connection, choice of 2-3 breathing circuits and induction chamber, user choice of multiple outputs while automatically adjusting for the correct flow rates. The induction chamber incorporates an automatic flush button to prevent undesired exposure when transferring animals. Includes individual timers for accurate monitoring and prevention of over exposure to the patients. Also includes all new digital warming beds to maintain body temperature during procedures, for both the induction chamber and breathing circuits between 95 to 104 degrees Fahrenheit.