Learn More
TARGETMOL CHEMICALS INC AD57 5MG
SDP
Catalog No. 502580944
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. AD57 is a potent inhibitor of both c-Src and Abl with IC50 of 0.025 uM and 0.041 uM respectively. purity: 98%
Catalog No. 50-258-0944 Supplier TARGETMOL CHEMICALS INC Supplier No. T22552L5MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. AD57 is a potent inhibitor of both c-Src and Abl with IC50 of 0.025 uM and 0.041 uM respectively. purity: 98%