Learn More
Cayman Chemical AD57 (hydrochloride), Purity: ≥98%, CAS: 2320261-72-9, 10 mg
SDP
Catalog No. 502954368
Encompass_Preferred
A polypharmacological cancer therapeutic; potently inhibits RET (IC50 = 2 nM) and reduces the activity of numerous other kinases by more than 80% when given at 1 μM in Drosophila; interferes with kinases downstream of RET, including Src, Raf, and S6K
Catalog No. 50-295-4368 Supplier Cayman Chemical Supplier No. 1397510 MG
A polypharmacological cancer therapeutic; potently inhibits RET (IC50 = 2 nM) and reduces the activity of numerous other kinases by more than 80% when given at 1 μM in Drosophila; interferes with kinases downstream of RET, including Src, Raf, and S6K