Learn More
Bioworld ADA (N-(2-acetamido)iminodiacetic acid) Buffer, 0.2M, pH 6.5, 250 mL
Catalog No. 50255328
Encompass_Preferred
Synonym:ADA-Na,N-(2-Acetamido)iminodiacetic acid monosodium salt,N-(Carbamoylmethyl)iminodiacetic acid monosodium saltpH: useful range: 6.0 - 7.2Warning: For laboratory use only.Not fit for agricultural, household, clinical use or human consumption. PH Value: : 6.0 - 7.2
Catalog No. 50-255-328 Supplier Bioworld Supplier No. 401200112
Synonym:ADA-Na,N-(2-Acetamido)iminodiacetic acid monosodium salt,N-(Carbamoylmethyl)iminodiacetic acid monosodium saltpH: useful range: 6.0 - 7.2Warning: For laboratory use only.Not fit for agricultural, household, clinical use or human consumption. PH Value: : 6.0 - 7.2