Learn More
Bioworld ADA (N-(2-acetamido)iminodiacetic acid) Buffer, 0.2M, pH 7.5, 250 mL
Catalog No. 50255334
Encompass_Preferred
Synonym:ADA-Na,N-(2-Acetamido)iminodiacetic acid monosodium salt,N-(Carbamoylmethyl)iminodiacetic acid monosodium saltA zwitterionic buffer used in biological applications including protein isolation and characterizationWarning: For laboratory use only.Not fit for agricultural, household, clinical use or human consumption. Appearance: LiquidPH Value: 7.5
Catalog No. 50-255-334 Supplier Bioworld Supplier No. 401200142
Synonym:ADA-Na,N-(2-Acetamido)iminodiacetic acid monosodium salt,N-(Carbamoylmethyl)iminodiacetic acid monosodium saltA zwitterionic buffer used in biological applications including protein isolation and characterizationWarning: For laboratory use only.Not fit for agricultural, household, clinical use or human consumption. Appearance: LiquidPH Value: 7.5