Learn More
Sigma Aldrich Fine Chemicals Biosciences ADA 98 titration1KG
Catalog No. 502907998
Encompass
ADA is a zwitterionic buffer used in biochemistry and molecular biology. It is one of the Good buffers developed in the 1960 to provide buffers in the pH range of 6.15 - 8.35 for wide applicability to biochemical studies. The pioneering publication by Good and co-workers describes the synthesis of ADA and its physical properties. The useful range of ADA buffer in aqueous SOLUTIONon is 6.0 - 7.2.
Catalog No. 50-290-7998 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. A98831KG
Add to Cart
Add to Cart
ADA is a zwitterionic buffer used in biochemistry and molecular biology. It is one of the Good buffers developed in the 1960 to provide buffers in the pH range of 6.15 - 8.35 for wide applicability to biochemical studies. The pioneering publication by Good and co-workers describes the synthesis of ADA and its physical properties. The useful range of ADA buffer in aqueous SOLUTIONon is 6.0 - 7.2.