Learn More
Medchemexpress LLC Ada disodium | 41689-31-0 | 98.0% | 10 G
SDP
Catalog No. 5000409971
Encompass_Preferred
ADA (disodium) is a buffer salt derived from ADA (adenosine deaminase) and is widely used in biochemical research, especially in the preparation of enzyme buffers. It is for research use only.
  • Buffer salt derived from adenosine deaminase
  • Widely used in biochemical research
  • Used in the preparation of enzyme buffers
Catalog No. 50-004-09971 Supplier Medchemexpress LLC Supplier No. HYD0855A10G
ADA (disodium) is a buffer salt derived from ADA (adenosine deaminase) and is widely used in biochemical research, especially in the preparation of enzyme buffers. It is for research use only.
  • Buffer salt derived from adenosine deaminase
  • Widely used in biochemical research
  • Used in the preparation of enzyme buffers