Learn More
Epigentek Inc ADA Polyclonal Antibody (100 ul)
SDP
Supplier:  Epigentek Inc A50009100
Encompass_Preferred
HRP Polyclonal Antibody Raised in Rabbit. Immunogen: Recombinant Human Adenosine deaminase protein (157-343AA). Tested Applications: ELISA. Reactivity: Human. Alternative Names: Adenosine deaminase, Adenosine aminohydrolase, ADA, ADA1.
Catalog No. 50-119-5976