Learn More
METTLER TOLEDO PTFE Eccentric Adapter, Robust Chemical Resistance, Compatible with 1/4 in Inserts, Quantity 1
Supplier:  METTLER TOLEDO 51162720
Encompass
Introducing the PTFE eccentric adapter, designed for advanced scientific applications in Mettler Toledo synthesis workstations. It provides exceptional chemical resistance and precise connectivity with an ST14/23 port size and a 6 mm inner diameter, compatible with 1/4 in (6.35 mm) inserts. With a durable PTFE GF25 construction, this adapter not only ensures secure, leak-free connections thanks to the included sealing ring but also stands out for its robust performance in demanding chemical environments.
Catalog No. NC1434336