Learn More
Enzo Life Sciences Ada-Tyr-(Ahx)3-(Leu)3-vinyl sulfone (100µg)
SDP
Catalog No. 502010331
Encompass_Preferred
A highly potent, cell-permeable proteasome inhibitor that targets all individual active subunits with comparable affinity. This extended peptide-based inhibitor can be radioiodinated, allowing detection of labeled subunits by autoradiography. Purity: ≥97%. Long Term Storage: -20°C.
Catalog No. 50-201-0331 Supplier Enzo Life Sciences Supplier No. BMLAW91600100
A highly potent, cell-permeable proteasome inhibitor that targets all individual active subunits with comparable affinity. This extended peptide-based inhibitor can be radioiodinated, allowing detection of labeled subunits by autoradiography. Purity: ≥97%. Long Term Storage: -20°C.