Learn More
Aviva Systems Biology Corp ADAD2 Antibody - middle region (ARP41114_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41114_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ADAD2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ADAD2. Alternate names: FLJ00337, TENRLApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-53275