Learn More
Medchemexpress LLC Adagrasib | 2326521-71-3 | >99.9% | 200 MG
SDP
Catalog No. 5000339616
Encompass_Preferred
Adagrasib (MRTX849) is a potent, orally-available, and mutation-selective covalent inhibitor of KRAS G12C with potential antineoplastic activity. It covalently binds to KRAS G12C at the cysteine at residue 12, locking the protein in its inactive GDP-bound conformation and inhibiting KRAS-dependent signal transduction.
  • Potent and orally-available.
  • Mutation-selective covalent inhibitor of KRAS G12C.
  • Potential antineoplastic activity.
  • Inhibits KRAS-dependent signal transduction.
Catalog No. 50-003-39616 Supplier Medchemexpress LLC Supplier No. HY130149200MG
Adagrasib (MRTX849) is a potent, orally-available, and mutation-selective covalent inhibitor of KRAS G12C with potential antineoplastic activity. It covalently binds to KRAS G12C at the cysteine at residue 12, locking the protein in its inactive GDP-bound conformation and inhibiting KRAS-dependent signal transduction.
  • Potent and orally-available.
  • Mutation-selective covalent inhibitor of KRAS G12C.
  • Potential antineoplastic activity.
  • Inhibits KRAS-dependent signal transduction.