Learn More
TARGETMOL CHEMICALS INC ADAGRASIB 25MG
SDP
Catalog No. 502579085
Encompass_Preferred
Also available in 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Adagrasib (MRTX849) is a potent selective and covalent KRASG12C inhibitor with potential antineoplastic activity. It selectively modifies mutant cysteine 12 in GDP-bound KRASG12C and inhibits KRAS-dependent signaling. purity: 99%
Catalog No. 50-257-9085 Supplier TARGETMOL CHEMICALS INC Supplier No. T836925MG
Also available in 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Adagrasib (MRTX849) is a potent selective and covalent KRASG12C inhibitor with potential antineoplastic activity. It selectively modifies mutant cysteine 12 in GDP-bound KRASG12C and inhibits KRAS-dependent signaling. purity: 99%