Learn More
Selleck Chemical LLC Adagrasib5mg
SDP
Supplier:  Selleck Chemical LLC S88845MG
Encompass_Preferred
Adagrasib (MRTX849) is a potent, selective, and covalent KRASG12C inhibitor that exhibits favorable drug-like properties, selectively modifies mutant cysteine 12 in GDP-bound KRASG12C and inhibits KRAS-dependent signaling. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3418