Learn More
ACROBiosystems ACROBiosystems Anti-Adalimumab Antibodies
Catalog No. 501957042
Encompass_Preferred
Antibody;100UG;IgG1/kappa;Recognizes Adalimumab specifically, no cross reactivity with other humanized antibodies.;Mouse monoclonal antibody is produced from a hybridoma resulting from fusion of SP2/0 myeloma and B-lymphocytes obtained from a mouse immunized with Adalimumab F(ab')2.; ;Anti-Adalimumab Antibodies bridging ELISA for Anti-Drug Antibody (ADA) assay development.Immobilized adalimumab at 1 µg/mL, add increasing concentrations of Anti-Adalimumab Antibodies (Cat. No. ADB-Y19, 10% human serum) and then add biotinylated adalimumab at 5 µg/mL. Detection was performed using HRP-conjugated streptavidin with a sensitivity of 0.6 ng/mL. ;Demonstration of the specificity of Anti-Adalimumab Antibodies (Cat. No. ADB-Y19) to the adalimumab.;Anti-Adalimumab Antibodies (mouse IgG1, Cat. No. ADB-Y19) captured on CM5 chip via anti-mouse antibodies surface, can bind human adalimumab with an affinity constant of 1.36 pM.
Catalog No. 50-195-7042 Supplier ACROBiosystems Supplier No. ADBY19100UG
Antibody;100UG;IgG1/kappa;Recognizes Adalimumab specifically, no cross reactivity with other humanized antibodies.;Mouse monoclonal antibody is produced from a hybridoma resulting from fusion of SP2/0 myeloma and B-lymphocytes obtained from a mouse immunized with Adalimumab F(ab')2.; ;Anti-Adalimumab Antibodies bridging ELISA for Anti-Drug Antibody (ADA) assay development.Immobilized adalimumab at 1 µg/mL, add increasing concentrations of Anti-Adalimumab Antibodies (Cat. No. ADB-Y19, 10% human serum) and then add biotinylated adalimumab at 5 µg/mL. Detection was performed using HRP-conjugated streptavidin with a sensitivity of 0.6 ng/mL. ;Demonstration of the specificity of Anti-Adalimumab Antibodies (Cat. No. ADB-Y19) to the adalimumab.;Anti-Adalimumab Antibodies (mouse IgG1, Cat. No. ADB-Y19) captured on CM5 chip via anti-mouse antibodies surface, can bind human adalimumab with an affinity constant of 1.36 pM.