Learn More
Eurofins Discoverx PathHunter Adalimumab Bioassay Kit, 10-Plate. (93-0538B15-00132)
Supplier:  Eurofins Discoverx 930538B1500132
Encompass
The PathHunter Adalimumab Bioassay Kit provides a robust, highly sensitive, and easy-to-use functional cell-based assay to measure drug potency and detect neutralizing antibodies. The bioassay kit contains all the materials needed to run the assay, including cryopreserved cells, cell plating reagent, control agonist, detection reagents, and assay plates. The ready-to-use cryopreserved cells have been manufactured for single-use to ensure assay reproducibility, and faster implementation from characterization to lot release. This assay has been optimized and qualified with Humira (not supplied in the bioassay kit). Humira is a registered trademark of AbbVie, Inc. For detailed product information, User manual and COA, visit www.discoverx.com/bioassay.
Catalog No. 50-200-0751
Add to Cart
Request a Quote