Learn More
Enzo Life Sciences ADAM 17 (CT) polyclonal antibody (100µg)
SDP
Supplier:  Enzo Life Sciences ADI905249100
Encompass_Preferred
Host: Rabbit. Immunogen: Synthetic peptide corresponding to the sequence near the C-terminus of human ADAM17. UniProt ID: P78536. Species reactivity: Human, Mouse, Rat. Applications: ICC, WB. Recommended Dilutions: Western Blot (1:500-1:2,000) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Peptide affinity purified. Formulation: Liquid. In PBS containing 0.02% sodium azide.
Catalog No. 50-200-6955