Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human ADAM11/MDC ELISA Kit

Catalog No. EHADAM11X5
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EHADAM11X5 5 x 96 Tests
EHADAM11 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. EHADAM11X5 Supplier Invitrogen™ Supplier No. EHADAM11X5
Get Up to $3,000 in Extra Life Sciences Consumables with Purchase
View all promotions
Add to Cart
Add to Cart

ELISA

The Human ADAM11/MDC ELISA quantitates Hu ADAM11 in human serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu ADAM11. Principle of the method The Human ADAM11 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

Probable ligand for integrin in the brain. This is a non catalytic metalloprotease-like protein.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O75078
Assay Range 1.5-400 pg/mL
Assay Sensitivity 1.5 pg/mL
Calibration Range 1.5 pg/mL to 400 pg/mL
Assay Solid-phase sandwich ELISA
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Protein Family Chemokines & Receptors
Sample Type Plasma, Serum, Supernatant
Specificity This ELISA kit shows no cross-reactivity with any of the cytokines tested: Human Angiogenin, BDNF, BLC, ENA-78, FGF-4, IL-1 alpha, IL-1 beta, IL-2, IL-3, IL-4, IL-5, IL-6, IL-7, IL-8, IL-9, IL-10, IL-11, IL- 12 p70, IL-12 p40, IL-13, IL-15, IL-309, IP-10, G-CSF, GM-CSF, IFN-gamma, Leptin, MCP-1, MCP-2, MCP-3, MIP-1 alpha, MIP-1 beta, MIP-1 delta, PARC, PDGF, RANTES, SCF, TARC, TGF-beta, TIMP-1, TIMP-2, TNF-alpha, TNF-beta, TPO, VEGF.
Sufficient For 5 x 96 Assays
Target Species Human
For Use With (Application) ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias a disintegrin and metallopeptidase domain 11; a disintegrin and metalloprotease domain (ADAM) 11; a disintegrin and metalloprotease domain 11; a disintegrin and metalloproteinase; ADAM; ADAM 11; ADAM metallopeptidase domain 11; Adam11; ADAMs; AW060611; disintegrin and metalloproteinase domain-containing protein 11; Mdc; metalloendopeptidases; metalloproteinase-like, disintegrin-like, and cysteine-rich proteina disintegrin and metallopeptidase domain 11; a disintegrin and metalloprotease domain (ADAM) 11; a disintegrin and metalloprotease domain 11; a disintegrin and metalloproteinase; ADAM; ADAM 11; ADAM metallopeptidase domain 11; Adam11; ADAMs; AW060611; disintegrin and metalloproteinase domain-containing protein 11; Mdc; metalloendopeptidases; metalloproteinase-like, disintegrin-like, and cysteine-rich protein; MDC
Gene ID (Entrez) 4185
Gene Symbol ADAM11
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Protein Aliases ADAM 11, Metalloproteinase-like, disintegrin-like, and cysteine-rich protein, MDC
Quantity 5 x 96 Tests
Regulatory Status RUO
Research Discipline Inflammation
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 100 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Synonym MDC
Target ADAM11
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.