Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human ADAM12 S Isoform Recombinant Protein

Catalog No. PIRP75521
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIRP75521 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIRP75521 Supplier Invitrogen™ Supplier No. RP75521
Only null left
Add to Cart
Add to Cart

Recombinant Protein

RP-75521 contains purified recombinant human ADAM12 S Isoform protein with an N-terminal hexahistidine tag. RP-75521 may be used as a positive control in many immunological assays.

TRUSTED_SUSTAINABILITY

Specifications

Accession Number O43184
Concentration 0.05 mg/mL
For Use With (Application) Control, Western Blot
Formulation Laemmli with no preservative; pH 6.8
Gene ID (Entrez) 8038
Name Human ADAM12 S Isoform
pH Range 6.8
Quantity 10 μg
Source E. Coli
Regulatory Status RUO
Gene Alias a disintegrin and metalloproteinase; ADAM; ADAM 12; ADAM metallopeptidase domain 12; ADAM12; ADAM12-OT1; ADAMs; CAR10; Disintegrin and metalloproteinase domain-containing protein 12; MCMP; MCMPMltna; meltrin-alpha; metalloendopeptidases; metalloprotease-disintegrin 12 transmembrane; MLTN; MLTNA; UNQ346/PRO545
Common Name ADAM12 S Isoform
Gene Symbol ADAM12
Conjugate Unconjugated
Species E. coli
Recombinant Recombinant
Protein Tag His-tag
Sequence Amino acids Glu208- Ile738 containing an N-terminal His-tag
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Protein Subtype Recombinant
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.