Learn More
Enzo Life Sciences ADAM17 fluorometric drug discovery kit (1x96wells)
SDP
Catalog No. 502010260
Encompass_Preferred
The ADAM17 Fluorometric (also known as fluorimetric) Drug Discovery Kit is a complete assay system designed to screen ADAM17 (TACE) inhibitors using a quenched fluorogenic peptide: Mca-PLAQAV-Dpa-RSSSR-NH2. Mca fluorescence is quenched by the Dpa group until cleavage by proteases separates the two moieties. The assays are performed in a convenient 96-well microplate format.  The kit is useful to screen inhibitors of ADAM17, a potential therapeutic target. The compound GM6001 is also included as a prototypic control inhibitor. Alternative name: TACE, A disintegrin and metalloproteinase 17. Applications: Fluorescent detection, HTS Activity assay.
Catalog No. 50-201-0260 Supplier Enzo Life Sciences Supplier No. BMLAK3100001
The ADAM17 Fluorometric (also known as fluorimetric) Drug Discovery Kit is a complete assay system designed to screen ADAM17 (TACE) inhibitors using a quenched fluorogenic peptide: Mca-PLAQAV-Dpa-RSSSR-NH2. Mca fluorescence is quenched by the Dpa group until cleavage by proteases separates the two moieties. The assays are performed in a convenient 96-well microplate format.  The kit is useful to screen inhibitors of ADAM17, a potential therapeutic target. The compound GM6001 is also included as a prototypic control inhibitor. Alternative name: TACE, A disintegrin and metalloproteinase 17. Applications: Fluorescent detection, HTS Activity assay.