Learn More
ACROBiosystems Recombinant Protein;Mouse ADAM17 / TACE / CD156b Protein;HEK293;500UG;AD7,ADAM17,TACE,CD156b
Catalog No. 501658770
Encompass_Preferred
Recombinant Protein;Mouse ADAM17 / TACE / CD156b Protein;500UG;Mouse ADAM17, His Tag (AD7-M52H1) is expressed from human 293 cells (HEK293). It contains AA Pro 18 - Asp 563 (Accession # Q9Z0F8).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 63.4 kDa. The protein migrates as 19 kDa and 20 kDa(Propeptide),55-66 kDa (Mature-ADAM17) and 75-94 kDa(Pro-ADAM17) under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-8770 Supplier ACROBiosystems Supplier No. AD7M52H1500UG
Recombinant Protein;Mouse ADAM17 / TACE / CD156b Protein;500UG;Mouse ADAM17, His Tag (AD7-M52H1) is expressed from human 293 cells (HEK293). It contains AA Pro 18 - Asp 563 (Accession # Q9Z0F8).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 63.4 kDa. The protein migrates as 19 kDa and 20 kDa(Propeptide),55-66 kDa (Mature-ADAM17) and 75-94 kDa(Pro-ADAM17) under reducing (R) condition (SDS-PAGE) due to glycosylation.