Learn More
Aviva Systems Biology Corp ADAM19 Antibody - N-terminal region (ARP49780_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49780_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ADAM19. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ADAM19. Alternate names: FKSG34, MADDAM, MLTNBApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-60179