Learn More
Aviva Systems Biology Corp ADAM30 Antibody - N-terminal region (ARP49661_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49661_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against ADAM30. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ADAM30. Alternate names: svph4ApplicationWB. Species reactivity: Cow, Dog, Human
Catalog No. 50-014-60140