Learn More
Aviva Systems Biology Corp ADAM33 Antibody - middle region (ARP49937_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49937_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ADAM33. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ADAM33. Alternate names: DJ964F7.1, DKFZp434K0521, FLJ35308, FLJ36751, MGC149823, MGC71889, C20orf153ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-60221