Learn More
ACROBiosystems Recombinant Protein;Human ADAM8 / CD156a Protein;HEK293;50UG;AD8,ADAM8,MS2,CD156a,CD156,ADAM-8,MS-2,ADAM
Supplier:  ACROBiosystems AD8H522350UG
Encompass_Preferred
Recombinant Protein;Human ADAM8 / CD156a Protein;50UG;Human ADAM8, His Tag (AD8-H5223) is expressed from human 293 cells (HEK293). It contains AA Ile 17 - Pro 497 (Accession # AAI15405.1).;rh ADAM8 fused with a polyhistidine tag at the C-terminus, has a calculated MW of 71.6 kDa, which migrates as a 120-150 kDa band in DTT-reduced SDS-PAGE due to glycosylation. Two main fragments bands are visible with apparent MW of 67 kDa and 46 kDa respectively in reduced SDS-PAGE as a result of its being prone to proteolytic cleavage.
Catalog No. 50-165-9071