Learn More
Macherey-Nagel TLC glass plates ADAMANT UV254, 0.25 mm, 10x10 cm
Catalog No. NC0951705
Encompass_Preferred
ADAMANT TLC precoated plates UV 254 - thickness of layer: 0.25 mm - size: 10x10 cm - pack of 25
Catalog No. NC0951705 Supplier Macherey-Nagel Supplier No. 821020
ADAMANT TLC precoated plates UV 254 - thickness of layer: 0.25 mm - size: 10x10 cm - pack of 25