Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Adamantane-1,3,5-triol | 99181-50-7 | MFCD08276262 | 184.24 | C10H16O3 | 25 G
SDP

Catalog No. 5000302757
Encompass_Preferred

Adamantane-1,3,5-triol is a trihydroxylated adamantane compound used as a drug intermediate in the synthesis of various active pharmaceutical and research compounds. Supplied for research use, it is handled as a solid and stored at low temperatures to preserve stability.

  • CAS number 99181-50-7.
  • Molecular formula C10H16O3.
  • Molecular weight 184.24 g/mol.
  • Appearance powder.
  • Storage: powder -20°C for 3 years, 4°C for 2 years; in solvent -80°C for 6 months, -20°C for 1 month.
  • Intended use: drug intermediate for synthesis of various active compounds.
  • Purity not stated on manufacturer datasheet.

Catalog No. 50-003-02757 Supplier Medchemexpress LLC Supplier No. 25GHYW016287
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Adamantane-1,3,5-triol is a trihydroxylated adamantane compound used as a drug intermediate in the synthesis of various active pharmaceutical and research compounds. Supplied for research use, it is handled as a solid and stored at low temperatures to preserve stability.

  • CAS number 99181-50-7.
  • Molecular formula C10H16O3.
  • Molecular weight 184.24 g/mol.
  • Appearance powder.
  • Storage: powder -20°C for 3 years, 4°C for 2 years; in solvent -80°C for 6 months, -20°C for 1 month.
  • Intended use: drug intermediate for synthesis of various active compounds.
  • Purity not stated on manufacturer datasheet.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.