Learn More
Medchemexpress LLC Adamantane-1,3,5-triol | 99181-50-7 | MFCD08276262 | 97.8% | 184.24 | C10H16O3 | 250 MG
SDP
Catalog No. 5000302783
Encompass_Preferred
Adamantane-1,3,5-triol is a small-molecule research chemical and drug intermediate (CAS 99181-50-7) used in organic synthesis and medicinal chemistry. Supplied as a high-purity solid, it functions as a versatile building block for synthesis of active compounds and laboratory research applications.
  • High purity (97.75%) suitable for research applications.
  • Solid powder form for easy handling and weighing.
  • Useful intermediate for organic synthesis and medicinal chemistry.
  • Stable under recommended storage conditions (powder: -20°C up to 3 years, 4°C up to 2 years).
  • Available in small pack sizes for laboratory-scale work.
Catalog No. 50-003-02783 Supplier Medchemexpress LLC Supplier No. 250MGHYW016287
Adamantane-1,3,5-triol is a small-molecule research chemical and drug intermediate (CAS 99181-50-7) used in organic synthesis and medicinal chemistry. Supplied as a high-purity solid, it functions as a versatile building block for synthesis of active compounds and laboratory research applications.
  • High purity (97.75%) suitable for research applications.
  • Solid powder form for easy handling and weighing.
  • Useful intermediate for organic synthesis and medicinal chemistry.
  • Stable under recommended storage conditions (powder: -20°C up to 3 years, 4°C up to 2 years).
  • Available in small pack sizes for laboratory-scale work.