Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Adamantane-1,3,5-triol | 99181-50-7 | MFCD08276262 | 97.8% | 184.24 g/mol | C10H16O3 | 50 G
SDP

Catalog No. 5000302183
Encompass_Preferred

Adamantane-1,3,5-triol is an adamantane-derived triol used as a synthetic intermediate in medicinal chemistry and related research applications. It is supplied as a powder intended for laboratory synthesis and small-scale research use.

  • Molecular formula: C10H16O3.
  • Molecular weight: 184.24 g/mol.
  • Physical appearance: powder.
  • Purity: 97.8% (HPLC) as reported on the certificate of analysis.
  • Storage: powder stable at -20°C (up to 3 years) and 4°C (up to 2 years); in solvent store at -80°C for longer-term or -20°C for short-term.
  • Application: used as a drug intermediate for synthesis of active compounds in research settings.

Catalog No. 50-003-02183 Supplier Medchemexpress LLC Supplier No. HYW01628750G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Adamantane-1,3,5-triol is an adamantane-derived triol used as a synthetic intermediate in medicinal chemistry and related research applications. It is supplied as a powder intended for laboratory synthesis and small-scale research use.

  • Molecular formula: C10H16O3.
  • Molecular weight: 184.24 g/mol.
  • Physical appearance: powder.
  • Purity: 97.8% (HPLC) as reported on the certificate of analysis.
  • Storage: powder stable at -20°C (up to 3 years) and 4°C (up to 2 years); in solvent store at -80°C for longer-term or -20°C for short-term.
  • Application: used as a drug intermediate for synthesis of active compounds in research settings.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.