Learn More
Medchemexpress LLC Adamantane-1,3,5-triol | 99181-50-7 | MFCD08276262 | 99.1% | 184.24 g/mol | C10H16O3 | 5 G
SDP
Catalog No. 5000302184
Encompass_Preferred
Adamantane-1,3,5-triol is a polycyclic cage compound employed as a synthetic intermediate in pharmaceutical and organic chemistry. The triol derivative provides a rigid, lipophilic scaffold (C10H16O3, MW 184.24 g/mol) useful for incorporating structural rigidity into target molecules during medicinal chemistry and materials research.
  • Serves as a drug intermediate for the synthesis of active compounds.
  • Provides a rigid, lipophilic cage scaffold for molecular design.
  • Characterized by formula C10H16O3 and molecular weight 184.24 g/mol.
  • Reported high purity suitable for synthetic chemistry applications.
  • Offered in multiple pack sizes to support small-scale and preparative work.
  • Stable when stored sealed and protected from moisture and light.
Catalog No. 50-003-02184 Supplier Medchemexpress LLC Supplier No. HYW0162875G
Adamantane-1,3,5-triol is a polycyclic cage compound employed as a synthetic intermediate in pharmaceutical and organic chemistry. The triol derivative provides a rigid, lipophilic scaffold (C10H16O3, MW 184.24 g/mol) useful for incorporating structural rigidity into target molecules during medicinal chemistry and materials research.
  • Serves as a drug intermediate for the synthesis of active compounds.
  • Provides a rigid, lipophilic cage scaffold for molecular design.
  • Characterized by formula C10H16O3 and molecular weight 184.24 g/mol.
  • Reported high purity suitable for synthetic chemistry applications.
  • Offered in multiple pack sizes to support small-scale and preparative work.
  • Stable when stored sealed and protected from moisture and light.