Learn More
Selleck Chemical LLC Adamantane S2869-100mg
SDP
Catalog No. 503112441
Encompass_Preferred
Adamantane is a colorless crystalline chemical compound first isolated from petroleum with a camphor-like odor Adamantane is used to treat influenza A virus infections
Catalog No. 50-311-2441 Supplier Selleck Chemical LLC Supplier No. S2869100MG
Adamantane is a colorless crystalline chemical compound first isolated from petroleum with a camphor-like odor Adamantane is used to treat influenza A virus infections