Learn More
Medchemexpress LLC Adamantane-d16 | 30470-60-1 | 100 MG
SDP
Catalog No. 5000420885
Encompass_Preferred
Adamantane-d16 is the deuterium labeled Adamantane. It is a polycyclic cage molecule with high symmetry and remarkable properties. It can be incorporated into a lipophilic part of the lipid bilayer that constitutes membranes and can act as an anchor in the lipid bilayer of liposomes.
  • Used as a tracer.
  • Used as an internal standard for quantitative analysis via NMR, GC-MS, or LC-MS.
  • Applications in research of surface recognition and drug delivery.
  • Incorporated as tracers for quantitation during drug development.
  • Deuteration can influence pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-20885 Supplier Medchemexpress LLC Supplier No. HYN2427S100MG
Adamantane-d16 is the deuterium labeled Adamantane. It is a polycyclic cage molecule with high symmetry and remarkable properties. It can be incorporated into a lipophilic part of the lipid bilayer that constitutes membranes and can act as an anchor in the lipid bilayer of liposomes.
  • Used as a tracer.
  • Used as an internal standard for quantitative analysis via NMR, GC-MS, or LC-MS.
  • Applications in research of surface recognition and drug delivery.
  • Incorporated as tracers for quantitation during drug development.
  • Deuteration can influence pharmacokinetic and metabolic profiles of drugs.