Learn More
Medchemexpress LLC Adamantane-d16 | 30470-60-1 | 25 MG
SDP
Catalog No. 5000421986
Encompass_Preferred
Adamantane-d16 is a deuterium-labeled polycyclic cage molecule known for its high symmetry and remarkable properties. It can be incorporated into the lipophilic part of lipid bilayers, serving as an anchor in liposomes. This compound is valuable in research for surface recognition and drug delivery studies. Deuteration of drug molecules, such as this, is also being studied for its potential to influence pharmacokinetic and metabolic profiles.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-21986 Supplier Medchemexpress LLC Supplier No. 25MGHYN2427S
Adamantane-d16 is a deuterium-labeled polycyclic cage molecule known for its high symmetry and remarkable properties. It can be incorporated into the lipophilic part of lipid bilayers, serving as an anchor in liposomes. This compound is valuable in research for surface recognition and drug delivery studies. Deuteration of drug molecules, such as this, is also being studied for its potential to influence pharmacokinetic and metabolic profiles.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS