Learn More
Medchemexpress LLC Adamantane-d16 | 30470-60-1 | 5 MG
SDP
Catalog No. 5000421883
Encompass_Preferred
Adamantane-d16 is the deuterium labeled Adamantane. Adamantane is a polycyclic cage molecule with high symmetry and remarkable properties. It can be incorporated into a lipophilic part of the lipid bilayer that constitutes membranes and can also serve as an anchor in the lipid bilayer of liposomes. It may be used in research of surface recognition and drug delivery. Deuteration has gained attention for its potential to affect the pharmacokinetic and metabolic profiles of drugs by incorporating stable heavy isotopes into drug molecules for quantitation during the drug development process.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-21883 Supplier Medchemexpress LLC Supplier No. HYN2427S5MG
Adamantane-d16 is the deuterium labeled Adamantane. Adamantane is a polycyclic cage molecule with high symmetry and remarkable properties. It can be incorporated into a lipophilic part of the lipid bilayer that constitutes membranes and can also serve as an anchor in the lipid bilayer of liposomes. It may be used in research of surface recognition and drug delivery. Deuteration has gained attention for its potential to affect the pharmacokinetic and metabolic profiles of drugs by incorporating stable heavy isotopes into drug molecules for quantitation during the drug development process.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS