Learn More
Medchemexpress LLC Adamantane-d16 | 30470-60-1 | 50 MG
SDP
Catalog No. 5000421105
Encompass_Preferred
Adamantane-d16 is the deuterium-labeled version of Adamantane. Adamantane is a polycyclic cage molecule known for its high symmetry and distinctive properties. It can be incorporated into the lipophilic part of lipid bilayers that form membranes and used as an anchor in the lipid bilayer of liposomes.
  • Utilized in research for surface recognition and drug delivery.
  • Can be used as a tracer for quantitation during drug development.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration has potential to influence the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-21105 Supplier Medchemexpress LLC Supplier No. HYN2427S50MG
Adamantane-d16 is the deuterium-labeled version of Adamantane. Adamantane is a polycyclic cage molecule known for its high symmetry and distinctive properties. It can be incorporated into the lipophilic part of lipid bilayers that form membranes and used as an anchor in the lipid bilayer of liposomes.
  • Utilized in research for surface recognition and drug delivery.
  • Can be used as a tracer for quantitation during drug development.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration has potential to influence the pharmacokinetic and metabolic profiles of drugs.