Learn More
TA Instruments Adamantane Traceable Temperature Enthalpy Calibration Material, Reference Material, Traceable Temperature and Enthalpy Calibration, DSC Cells, Accurate Reliable Measurements
Catalog No. 5000580630 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments Adamantane Traceable Temperature Enthalpy Calibration Material is a reference material used for traceable temperature and enthalpy calibration of DSC cells, ensuring accurate and reliable DSC measurements.
Catalog No. 50-005-80630 Supplier TA Instruments Supplier No. 970489.901
Waters TA Instruments Adamantane Traceable Temperature Enthalpy Calibration Material is a reference material used for traceable temperature and enthalpy calibration of DSC cells, ensuring accurate and reliable DSC measurements.