Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Adamantanine (2-aminoadamantane-2-carboxylic acid) | 42381-05-5 | ≥97.0% | 250 MG
SDP

Catalog No. 5000233460
Encompass_Preferred

Adamantanine (2-aminoadamantane-2-carboxylic acid) is a small-molecule research compound used to probe amino acid transport and aminopeptidase-related pathways in cellular and biochemical studies.

  • CAS number 42381-05-5
  • Molecular formula C11H17NO2
  • Molecular weight 195.26 g/mol
  • Purity ≥97.0%
  • Appearance solid, off-white to yellow
  • Available in 50 mg-5 g packs; 10 g and 50 g available by quote
  • Store sealed, away from moisture and light; in solvent store at -80°C (6 months) or -20°C (1 month)

Catalog No. 50-002-33460 Supplier Medchemexpress LLC Supplier No. HY121123250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Adamantanine (2-aminoadamantane-2-carboxylic acid) is a small-molecule research compound used to probe amino acid transport and aminopeptidase-related pathways in cellular and biochemical studies.

  • CAS number 42381-05-5
  • Molecular formula C11H17NO2
  • Molecular weight 195.26 g/mol
  • Purity ≥97.0%
  • Appearance solid, off-white to yellow
  • Available in 50 mg-5 g packs; 10 g and 50 g available by quote
  • Store sealed, away from moisture and light; in solvent store at -80°C (6 months) or -20°C (1 month)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.