Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Arkray Adams HA 8180V Analyzer
SDP

Catalog No. 21377286
Change view
Click to view available options
Sample Type:
Urine
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Type
21-377-286 Urine
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
For professional in vitro diagnostic use only. Handle blood samples and reagents according to standard biosafety guidelines. Wear protective gloves and eyewear. Avoid ingestion or contact with skin and eyes. Dispose of biohazardous waste properly.
Catalog No. 21-377-286 Supplier Arkray Supplier No. 100675
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For professional in vitro diagnostic use only. Handle blood samples and reagents according to standard biosafety guidelines. Wear protective gloves and eyewear. Avoid ingestion or contact with skin and eyes. Dispose of biohazardous waste properly.

The Adams HA-8180V Analyzer delivers fast, precise HbA1c results with minimal operator intervention. The HA-8180V enhances workflow efficiency, improves patient turnaround, and offers reliable accuracy to support diabetes monitoring in any clinical lab.

  • Fully-automated HPLC glyco‐hemoglobin analyzer: The ADAMS A1c HA‐8180V uses high‐performance liquid chromatography (HPLC) – the gold standard for HbA1c measurement – ensuring highly accurate and reliable detection of glycohemoglobin levels in whole blood or hemolysate samples
  • Dual modes for flexible workflows (Fast Mode and Variant Mode):
    • Fast Mode: Provides rapid turnaround at roughly 48 seconds per test – ideal for high-throughput environments or routine monitoring. :contentReference[oaicite:2]{index=2}
    • Variant Mode: Processes samples in about 90 seconds per test and supports hemoglobin variant detection (e.g. HbS, HbC, and flags for HbD/HbE), enabling accurate HbA1c measurement even in patients with hemoglobin variants.
  • Broad measurement scope – HbA1c plus HbF detection: The unit measures stable HbA1c (S‐A1c) and fetal hemoglobin (HbF), and in Variant Mode can detect variants such as HbS and HbC (plus flagging HbD/HbE), helping avoid misinterpretation of HbA1c due to variant interference.
  • Exceptional precision and accuracy: The analyzer delivers low coefficient of variation (CV ≤ 1%), giving clinicians confidence in the reliability and reproducibility of results – critical for diagnosis and long‐term monitoring of diabetes.
  • Low sample volume requirement: Only ~14 μL of whole blood is needed per test, minimizing patient burden and allowing efficient use of scarce samples.
  • High throughput and capacity: The system can process up to roughly 37 tests/hour, making it suitable for busy labs. It supports sample racks with one‐way (50 samples) or loop (100 samples) loading, enabling continuous, walk‐away operation.
  • User‐friendly design and automation reduces workload:
    • Cap‐piercing sampler: allows capped blood collection tubes to be loaded directly, eliminating manual sample preparation and improving biosafety.
    • Large color LCD display: shows reagent levels and chromatogram graphs, providing intuitive monitoring and status feedback.
    • Built‐in thermal printer (58 mm width): enables on‐the‐spot printing of results for record‐keeping or reporting.
    • Memory capacity: can store up to 900 measurement results (including calibration data), facilitating data management and traceability.
    • Flexible connectivity: supports RS‐232C serial communication (bi‐directional) or Ethernet (optional), enabling integration with lab information systems (LIS).
  • Minimal maintenance and lower operating cost: The integrated column design eliminates the need for frequent pre-filter changes. Combined with efficient aluminium reagent supply, this reduces reagent and maintenance costs while ensuring consistent performance.
  • Compact, benchtop footprint suitable for clinical labs: The unit measures 530 mm x 530 mm x 530 mm and weighs roughly 39 kg (plus ~4 kg for the sampler) – compact enough for typical lab benches while still offering full automation and high performance.
  • Regulatory clearance and clinical diagnostic use: The HA‐8180V is cleared (FDA 510(k)) for use as an aid in the diagnosis and monitoring of diabetes mellitus – making it suitable for professional diagnostic labs.
  • Reliable long-term glucose monitoring support: By measuring HbA1c – a widely accepted marker for long-term glycemic control – the analyzer helps clinicians track patient blood glucose over time, guide treatment, and assess risk of diabetes-related complications.

Specifications

Type Hemoglobin Analyzer
Sample Type Urine

The ADAMS A1c HA‐8180V Analyzer offers rapid, fully automated HbA1c testing with variant detection, designed to streamline workflows in busy clinical laboratories.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.