Learn More
Medchemexpress LLC ADAMTS-5 Inhibitor 50mg | 929634-33-3 | 449.92 g·mol⁻¹ | C16H11ClF3N3OS3 | 50 MG
SDP
Catalog No. 5000227724
Encompass_Preferred
ADAMTS-5 Inhibitor is a small-molecule research compound that potently inhibits ADAMTS-5 (aggrecanase-2) with an IC50 of 1.1 μM and shows >40-fold functional selectivity over ADAMTS-4. It is supplied as a solid with a reported purity of 99.59%, a molecular weight of 449.92 g·mol⁻¹, and formula C16H11ClF3N3OS3. For research use only.
  • Potent ADAMTS-5 inhibition (IC50 1.1 μM).
  • Greater than 40-fold functional selectivity over ADAMTS-4.
  • High reported purity of 99.59%.
  • Supplied as a solid; also available as a 10 mM solution in DMSO.
  • Available in multiple milligram packaging options (5 mg-500 mg) and larger quantities on request.
  • Intended for research use only.
Catalog No. 50-002-27724 Supplier Medchemexpress LLC Supplier No. HY11499650MG
ADAMTS-5 Inhibitor is a small-molecule research compound that potently inhibits ADAMTS-5 (aggrecanase-2) with an IC50 of 1.1 μM and shows >40-fold functional selectivity over ADAMTS-4. It is supplied as a solid with a reported purity of 99.59%, a molecular weight of 449.92 g·mol⁻¹, and formula C16H11ClF3N3OS3. For research use only.
  • Potent ADAMTS-5 inhibition (IC50 1.1 μM).
  • Greater than 40-fold functional selectivity over ADAMTS-4.
  • High reported purity of 99.59%.
  • Supplied as a solid; also available as a 10 mM solution in DMSO.
  • Available in multiple milligram packaging options (5 mg-500 mg) and larger quantities on request.
  • Intended for research use only.